siRNA Details
| Gene Symbol: Clec1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175526.3 | NP_780735.2 | 1 | 328 | ||
| GenBank mRNA | AK031121.1 | - | 1 | 150 | ||
| AK036846.1 | - | 1 | 183 | |||
| AK049608.1 | - | 1 | 75 | |||
| AK165920.1 | - | 1 | 187 | |||
| BC052840.1 | - | 1 | 187 | 30050488 | ||
| BX512526.1 | - | 1 | 328 | |||

Clec1a
Genomic Map