siRNA Details
| Gene Symbol: Fkrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173430.2 | NP_775606.1 | 3 | 963 | ||
| GenBank mRNA | AJ511806.1 | - | 3 | 946 | ||
| AK080624.1 | - | 1 | 773 | |||
| AK135089.1 | - | 3 | 963 | |||
| AK156156.1 | - | 3 | 962 | |||
| AK166068.1 | - | 3 | 965 | |||
| AK168308.1 | - | 3 | 961 | |||
| BC053072.1 | - | 3 | 946 | 5686286 | ||

Fkrp
Genomic Map