siRNA Details
| Gene Symbol: Nlrp9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194058.2 | NP_918947.2 | 8 | 2766 | ||
| GenBank mRNA | AK135777.1 | - | 5 | 898 | ||
| AY596196.1 | - | 8 | 2766 | |||
| BC141249.1 | - | 8 | 2764 | 9055861 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrp9b| Gene Symbol: Nlrp9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194058.2 | NP_918947.2 | 8 | 2766 | ||
| GenBank mRNA | AK135777.1 | - | 5 | 898 | ||
| AY596196.1 | - | 8 | 2766 | |||
| BC141249.1 | - | 8 | 2764 | 9055861 | ||