siRNA Details
| Gene Symbol: Ggn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182694.2 | NP_874353.2 | 3 | 2570 | ||
| NM_182696.2 | NP_874355.1 | 3 | 526 | |||
| GenBank mRNA | AF538032.1 | - | 3 | 2059 | ||
| AF538033.1 | - | 4 | 1479 | |||
| AF538034.1 | - | 3 | 526 | |||
| BC089358.1 | - | 3 | 2570 | 30610755 | ||

Ggn
Genomic Map