siRNA Details
Gene Symbol: Cd163l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172909.4 | NP_766497.3 | 8 | 2535 | ||
GenBank mRNA | AF539744.1 | - | 8 | 2291 | ||
AK046675.1 | - | 7 | 2659 | |||
AK088010.1 | - | 8 | 2522 | |||
BC145639.1 | - | 7 | 2384 | |||
BC145640.1 | - | 7 | 2384 |
