siRNA Details
| Gene Symbol: Cd163l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172909.4 | NP_766497.3 | 12 | 3001 | ||
| GenBank mRNA | AF539744.1 | - | 12 | 2757 | ||
| AK046675.1 | - | 11 | 3178 | |||
| AK088010.1 | - | 11 | 2994 | |||
| AW494409.1 | - | 3 | 465 | |||
| BC145639.1 | - | 10 | 2826 | |||
| BC145640.1 | - | 11 | 2850 | |||
| BF469445.1 | - | 3 | 231 | |||

Cd163l1
Genomic Map