siRNA Details
Gene Symbol: Mrgpre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175534.3 | NP_780743.1 | 2 | 618 | ||
GenBank mRNA | AJ320231.1 | - | 2 | 329 | ||
AK048528.1 | - | 2 | 436 | |||
AK133657.1 | - | 2 | 620 | |||
AK154886.1 | - | 2 | 413 | |||
BC096766.1 | - | 2 | 414 | 30102203 | ||
BC138305.1 | - | 2 | 327 | 8861325 | ||
BC138308.1 | - | 2 | 327 | 8861328 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
