siRNA Details
| Gene Symbol: Elmod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170691.1 | NP_001164162.1 | 9 | 982 | ||
| NM_178736.5 | NP_848851.3 | 9 | 987 | |||
| GenBank mRNA | AK029044.1 | - | 9 | 958 | ||
| AK036607.1 | - | 9 | 966 | |||
| AK036746.1 | - | 9 | 927 | |||
| AK041137.1 | - | 9 | 888 | |||
| AK048572.1 | - | 5 | 2193 | |||
| AK082730.1 | - | 8 | 754 | |||
| AK084950.1 | - | 9 | 987 | |||
| BC079654.1 | - | 9 | 948 | 6855187 | ||

Elmod2
Genomic Map