siRNA Details
Gene Symbol: Layn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033534.1 | NP_001028706.1 | 7 | 891 | ||
GenBank mRNA | BC139816.1 | - | 863 | 40140447 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Layn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033534.1 | NP_001028706.1 | 7 | 891 | ||
GenBank mRNA | BC139816.1 | - | 863 | 40140447 |