siRNA Details
| Gene Symbol: Zc3h12c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162921.1 | NP_001156393.1 | 6 | 2288 | ||
| GenBank mRNA | AK220416.1 | - | 4 | 1418 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h12c| Gene Symbol: Zc3h12c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162921.1 | NP_001156393.1 | 6 | 2288 | ||
| GenBank mRNA | AK220416.1 | - | 4 | 1418 | ||