siRNA Details
| Gene Symbol: Scaper | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081341.1 | NP_001074810.1 | 16, 17 | 2047 | ||
| GenBank mRNA | AK049497.1 | - | 16, 17 | 2043 | ||
| AK173175.1 | - | 15, 16 | 1977 | |||
| BC095992.1 | - | 15, 16 | 1977 | |||
| BC157959.1 | - | 15, 16 | 1992 | 9007286 | ||
| BC158062.1 | - | 15, 16 | 1969 | 9088165 | ||
| BC172119.1 | - | 15, 16 | 1992 | 9054405 | ||
| BC172151.1 | - | 15, 16 | 1969 | 9054821 | ||

Scaper
Genomic Map