siRNA Details
| Gene Symbol: Scaper | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081341.1 | NP_001074810.1 | 22 | 2725 | ||
| GenBank mRNA | AK048755.1 | - | 11 | 1253 | ||
| AK049497.1 | - | 22 | 2721 | |||
| AK173175.1 | - | 21 | 2655 | |||
| BC095992.1 | - | 21 | 2655 | |||
| BC157959.1 | - | 21 | 2670 | 9007286 | ||
| BC158062.1 | - | 21 | 2647 | 9088165 | ||
| BC172119.1 | - | 21 | 2670 | 9054405 | ||
| BC172151.1 | - | 21 | 2647 | 9054821 | ||
| EF410111.1 | - | 939 | ||||

Scaper
Genomic Map