siRNA Details
| Gene Symbol: Prss35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178738.3 | NP_848853.2 | 2 | 483 | ||
| GenBank mRNA | AK030671.1 | - | 2 | 468 | ||
| AK031411.1 | - | 2 | 468 | |||
| AK031644.1 | - | 2 | 468 | |||
| AK076281.1 | - | 2 | 468 | |||
| BC075675.1 | - | 2 | 452 | 30547401 | ||
| DQ223037.1 | - | 2 | 443 | |||

Prss35
Genomic Map