siRNA Details
| Gene Symbol: Dclk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172928.5 | NP_766516.2 | 5 | 2524 | ||
| GenBank mRNA | AK050312.1 | - | 5 | 2492 | ||
| AK135010.1 | - | 5 | 2513 | |||
| BC056929.1 | - | 4 | 2192 | 6816292 | ||
| DQ286388.1 | - | 5 | 2493 | |||

Dclk3
Genomic Map