siRNA Details
| Gene Symbol: Gm10605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK141001.1 | - | 1 | 105 | ||
| Gene Symbol: Gm4980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195529.1 | NP_001182458.1 | 1 | 1061 | ||
| GenBank mRNA | AK032609.1 | - | 1 | 134 | ||

Gm10605
Genomic Map