siRNA Details
| Gene Symbol: 9930111J21Rik2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173434.1 | NP_775610.1 | 3 | 1657 | ||
| GenBank mRNA | AK037088.1 | - | 3 | 1657 | ||
| BC119805.1 | - | 2 | 1561 | 40107440 | ||
| BC122882.1 | - | 2 | 1566 | 40107441 | ||

9930111J21Rik2
Genomic Map