siRNA Details
Gene Symbol: E330021D16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201390.1 | NP_001188319.1 | 2 | 521 | ||
GenBank mRNA | AK054382.1 | - | 2 | 522 | ||
AK135843.1 | - | 2 | 522 | |||
AK162229.1 | - | 2 | 451 | |||
BC099944.1 | - | 2 | 512 |
Gene Symbol: AA414768 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272033.1 | NP_001258962.1 | 1 | 311 | ||
GenBank mRNA | CJ053549.1 | - | 1 | 310 | ||
CJ131333.1 | - | 1 | 311 |
