siRNA Details
| Gene Symbol: Dcaf12l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175539.3 | NP_780748.1 | 1 | 1013 | ||
| GenBank mRNA | AK037333.1 | - | 2 | 963 | ||
| AK076436.1 | - | 1020 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcaf12l2| Gene Symbol: Dcaf12l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175539.3 | NP_780748.1 | 1 | 1013 | ||
| GenBank mRNA | AK037333.1 | - | 2 | 963 | ||
| AK076436.1 | - | 1020 | ||||