siRNA Details
| Gene Symbol: Cypt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173436.2 | NP_775612.2 | 2 | 548 | ||
| GenBank mRNA | AF463499.1 | - | 2 | 531 | ||
| BC060953.1 | - | 2 | 548 | 6774045 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cypt2| Gene Symbol: Cypt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173436.2 | NP_775612.2 | 2 | 548 | ||
| GenBank mRNA | AF463499.1 | - | 2 | 531 | ||
| BC060953.1 | - | 2 | 548 | 6774045 | ||