siRNA Details
| Gene Symbol: Pcdh11x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271809.1 | NP_001258738.1 | 3 | 1608 | ||
| NM_001271810.1 | NP_001258739.1 | 3 | 1608 | |||
| GenBank mRNA | AF383875.1 | - | 1 | 27 | ||
| AJ564976.1 | - | 3 | 1675 | |||
| AK039071.1 | - | 3 | 1609 | |||
| BC126930.1 | - | 3 | 1531 | 40086368 | ||
| EU854474.1 | - | 1 | 370 | |||

Pcdh11x
Genomic Map