siRNA Details
| Gene Symbol: Zfp711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177747.3 | NP_808415.2 | 8 | 1552 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp711| Gene Symbol: Zfp711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177747.3 | NP_808415.2 | 8 | 1552 | ||