siRNA Details
| Gene Symbol: Tbc1d8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081499.2 | NP_001074968.1 | 2 | 403 | ||
| GenBank mRNA | BC147581.1 | - | 2 | 266 | 9056571 | |
| BC151057.1 | - | 2 | 266 | 9087970 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d8b| Gene Symbol: Tbc1d8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081499.2 | NP_001074968.1 | 2 | 403 | ||
| GenBank mRNA | BC147581.1 | - | 2 | 266 | 9056571 | |
| BC151057.1 | - | 2 | 266 | 9087970 | ||