siRNA Details
Gene Symbol: Rbbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009031.3 | NP_033057.3 | 2 | 410 | ||
GenBank mRNA | AK076016.1 | - | 2 | 335 | ||
AK078349.1 | - | 2 | 323 | |||
AK082892.1 | - | 2 | 323 | |||
AK132101.1 | - | 2 | 320 | |||
AK135779.1 | - | 2 | 324 | |||
AK135869.1 | - | 1 | 19 | |||
AK135875.1 | - | 1 | 19 | |||
AK135956.1 | - | 2 | 324 | |||
AK136110.1 | - | 2 | 322 | |||
AK145531.1 | - | 2 | 321 | |||
AK145651.1 | - | 2 | 304 | |||
AK146014.1 | - | 2 | 322 | |||
AK146439.1 | - | 2 | 381 | |||
AK146904.1 | - | 2 | 323 | |||
AK146967.1 | - | 2 | 322 | |||
AK147062.1 | - | 2 | 381 | |||
AK148852.1 | - | 2 | 350 | |||
AK153913.1 | - | 2 | 367 | |||
AK160023.1 | - | 2 | 322 | |||
BC003785.1 | - | 2 | 339 | 3600013 | ||
U35142.1 | - | 2 | 349 |
