siRNA Details
| Gene Symbol: Atg9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003917.4 | NP_001003917.2 | 8 | 1203 | ||
| NM_001288612.1 | NP_001275541.1 | 8 | 1206 | |||
| NM_001288613.1 | NP_001275542.1 | 8 | 1451 | |||
| GenBank mRNA | AK210178.1 | - | 1 | 156 | ||
| AM085510.1 | - | 8 | 1116 | |||
| BC056488.1 | - | 8 | 1364 | |||
| BC079884.1 | - | 8 | 1151 | 6840050 | ||
| BC082307.1 | - | 6 | 909 | |||

Atg9a
Genomic Map