siRNA Details
| Gene Symbol: Atg9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003917.4 | NP_001003917.2 | 11, 12 | 2125 | ||
| NM_001288612.1 | NP_001275541.1 | 11, 12 | 2128 | |||
| NM_001288613.1 | NP_001275542.1 | 11, 12 | 2373 | |||
| GenBank mRNA | AM085510.1 | - | 11, 12 | 2038 | ||
| BC034139.1 | - | 4, 5 | 814 | |||
| BC056488.1 | - | 11, 12 | 2286 | |||
| BC079884.1 | - | 11, 12 | 2014 | 6840050 | ||
| BC082307.1 | - | 9, 10 | 1830 | |||

Atg9a
Genomic Map