siRNA Details
| Gene Symbol: Ift52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172150.4 | NP_742162.2 | 8 | 713 | ||
| GenBank mRNA | AK088467.1 | - | 8 | 714 | ||
| AK144188.1 | - | 8 | 813 | |||
| AK160745.1 | - | 8 | 718 | |||
| AK167194.1 | - | 8 | 729 | |||
| AK178241.1 | - | 2 | 80 | |||
| BC037708.1 | - | 8 | 682 | 4222723 | ||

Ift52
Genomic Map