siRNA Details
| Gene Symbol: Rhbdl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139228.3 | NP_631974.1 | 8 | 1178 | ||
| GenBank mRNA | AJ313479.1 | - | 8 | 1178 | ||
| BC054784.1 | - | 8 | 1014 | 6817213 | ||
| BC056969.1 | - | 8 | 1014 | 6817213 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhbdl3| Gene Symbol: Rhbdl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139228.3 | NP_631974.1 | 8 | 1178 | ||
| GenBank mRNA | AJ313479.1 | - | 8 | 1178 | ||
| BC054784.1 | - | 8 | 1014 | 6817213 | ||
| BC056969.1 | - | 8 | 1014 | 6817213 | ||