siRNA Details
Gene Symbol: Bivm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144558.4 | NP_653141.3 | 4, 5 | 1129 | ||
GenBank mRNA | AF411386.1 | - | 5, 6 | 1132 | ||
AK032375.1 | - | 4, 5 | 1089 | |||
AK034329.1 | - | 4, 5 | 1091 | |||
BC082584.1 | - | 4, 5 | 935 | 30612339 |
