siRNA Details
| Gene Symbol: Prok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044382.1 | NP_001037847.1 | 1 | 82 | ||
| GenBank mRNA | BC117006.1 | - | 1 | 82 | 40126325 | |
| BC117008.1 | - | 1 | 82 | 40126327 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prok1| Gene Symbol: Prok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044382.1 | NP_001037847.1 | 1 | 82 | ||
| GenBank mRNA | BC117006.1 | - | 1 | 82 | 40126325 | |
| BC117008.1 | - | 1 | 82 | 40126327 | ||