siRNA Details
| Gene Symbol: Rhobtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153514.5 | NP_705734.4 | 6, 7 | 2143 | ||
| GenBank mRNA | AF420001.1 | - | 5, 6 | 1634 | ||
| AK038196.1 | - | 6, 7 | 2235 | |||
| AK081100.1 | - | 6, 7 | 2072 | |||
| AK122354.1 | - | 6, 7 | 2124 | |||
| AK171295.1 | - | 12, 13 | 2758 | |||
| BC026836.1 | - | 6, 7 | 2124 | 4242008 | ||
| BC056954.1 | - | 6, 7 | 1748 | 5685456 | ||

Rhobtb2
Genomic Map