siRNA Details
| Gene Symbol: Dusp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145744.2 | NP_665687.1 | 6 | 341 | ||
| GenBank mRNA | AF357887.1 | - | 6 | 341 | ||
| BC116841.1 | - | 6 | 336 | 40126160 | ||
| BC116843.1 | - | 6 | 336 | 40126162 | ||
| BC144711.1 | - | 5 | 302 | |||

Dusp15
Genomic Map