siRNA Details
| Gene Symbol: Olfr1344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177061.3 | NP_796035.2 | 1 | 104 | ||
| GenBank mRNA | BC147050.1 | - | 1 | 170 | 9056072 | |
| BC147051.1 | - | 1 | 170 | 9056049 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1344| Gene Symbol: Olfr1344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177061.3 | NP_796035.2 | 1 | 104 | ||
| GenBank mRNA | BC147050.1 | - | 1 | 170 | 9056072 | |
| BC147051.1 | - | 1 | 170 | 9056049 | ||