siRNA Details
Gene Symbol: Olfr132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005481.1 | NP_001005481.1 | 1 | 612 | ||
GenBank mRNA | BC107196.2 | - | 1 | 611 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005481.1 | NP_001005481.1 | 1 | 612 | ||
GenBank mRNA | BC107196.2 | - | 1 | 611 |