siRNA Details
| Gene Symbol: Olfr115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011753.2 | NP_001011753.2 | 1 | 647 | ||
| GenBank mRNA | BC139226.1 | - | 1 | 647 | 8862248 | |
| BC139227.1 | - | 1 | 647 | 8862249 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr115| Gene Symbol: Olfr115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011753.2 | NP_001011753.2 | 1 | 647 | ||
| GenBank mRNA | BC139226.1 | - | 1 | 647 | 8862248 | |
| BC139227.1 | - | 1 | 647 | 8862249 | ||