siRNA Details
| Gene Symbol: Olfr1229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011761.1 | NP_001011761.1 | 1 | 142 | ||
| GenBank mRNA | BC147797.1 | - | 1 | 168 | 9087514 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1229| Gene Symbol: Olfr1229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011761.1 | NP_001011761.1 | 1 | 142 | ||
| GenBank mRNA | BC147797.1 | - | 1 | 168 | 9087514 | |