siRNA Details
Gene Symbol: Olfr1229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011761.1 | NP_001011761.1 | 1 | 452 | ||
GenBank mRNA | BC147797.1 | - | 1 | 478 | 9087514 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011761.1 | NP_001011761.1 | 1 | 452 | ||
GenBank mRNA | BC147797.1 | - | 1 | 478 | 9087514 |