siRNA Details
| Gene Symbol: Olfr1042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011777.2 | NP_001011777.1 | 1 | 313 | ||
| GenBank mRNA | BC150927.1 | - | 1 | 313 | 9087838 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1042| Gene Symbol: Olfr1042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011777.2 | NP_001011777.1 | 1 | 313 | ||
| GenBank mRNA | BC150927.1 | - | 1 | 313 | 9087838 | |