siRNA Details
| Gene Symbol: Olfr301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212436.2 | NP_997601.1 | 3 | 674 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr301| Gene Symbol: Olfr301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212436.2 | NP_997601.1 | 3 | 674 | ||