siRNA Details
| Gene Symbol: Olfr93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011813.1 | NP_001011813.1 | 1 | 438 | ||
| GenBank mRNA | BC139238.1 | - | 1 | 519 | 8862260 | |
| BC139240.1 | - | 1 | 519 | 8862262 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr93| Gene Symbol: Olfr93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011813.1 | NP_001011813.1 | 1 | 438 | ||
| GenBank mRNA | BC139238.1 | - | 1 | 519 | 8862260 | |
| BC139240.1 | - | 1 | 519 | 8862262 | ||