siRNA Details
| Gene Symbol: Olfr538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011867.1 | NP_001011867.1 | 1 | 63 | ||
| Gene Symbol: Olfr537-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC108946.1 | - | 1 | 87 | ||

Olfr538
Genomic Map