siRNA Details
| Gene Symbol: Olfr417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207137.3 | NP_997020.3 | 1 | 142 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr417| Gene Symbol: Olfr417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207137.3 | NP_997020.3 | 1 | 142 | ||
Set Membership: |
|