siRNA Details
| Gene Symbol: Olfr1324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146292.1 | NP_666404.1 | 1 | 490 | ||
| GenBank mRNA | BC119382.1 | - | 1 | 578 | 8734131 | |
| BC120627.1 | - | 1 | 578 | 40129550 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1324| Gene Symbol: Olfr1324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146292.1 | NP_666404.1 | 1 | 490 | ||
| GenBank mRNA | BC119382.1 | - | 1 | 578 | 8734131 | |
| BC120627.1 | - | 1 | 578 | 40129550 | ||