siRNA Details
Gene Symbol: Olfr827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146300.1 | NP_666412.1 | 1 | 165 | ||
GenBank mRNA | BC117109.1 | - | 1 | 186 | 40126428 | |
BC117111.1 | - | 1 | 186 | 40126430 |
