siRNA Details
| Gene Symbol: Olfr1449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146303.1 | NP_666415.1 | 1 | 141 | ||
| GenBank mRNA | BC133705.1 | - | 1 | 145 | 40135405 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1449| Gene Symbol: Olfr1449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146303.1 | NP_666415.1 | 1 | 141 | ||
| GenBank mRNA | BC133705.1 | - | 1 | 145 | 40135405 | |