siRNA Details
| Gene Symbol: Olfr187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146322.2 | NP_666434.2 | 2 | 774 | ||
| GenBank mRNA | BC119304.1 | - | 1 | 662 | 8734053 | |
| BC119306.1 | - | 1 | 665 | 8734055 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr187| Gene Symbol: Olfr187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146322.2 | NP_666434.2 | 2 | 774 | ||
| GenBank mRNA | BC119304.1 | - | 1 | 662 | 8734053 | |
| BC119306.1 | - | 1 | 665 | 8734055 | ||