siRNA Details
| Gene Symbol: Olfr1330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146334.2 | NP_666446.1 | 1 | 579 | ||
| GenBank mRNA | BC108943.1 | - | 1 | 579 | 40042936 | |
| BC108944.1 | - | 1 | 574 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1330| Gene Symbol: Olfr1330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146334.2 | NP_666446.1 | 1 | 579 | ||
| GenBank mRNA | BC108943.1 | - | 1 | 579 | 40042936 | |
| BC108944.1 | - | 1 | 574 | |||