siRNA Details
| Gene Symbol: Olfr374 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146338.2 | NP_666450.2 | 2 | 627 | ||
| GenBank mRNA | BC145662.1 | - | 1 | 571 | 40130965 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr374| Gene Symbol: Olfr374 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146338.2 | NP_666450.2 | 2 | 627 | ||
| GenBank mRNA | BC145662.1 | - | 1 | 571 | 40130965 | |