siRNA Details
Gene Symbol: Olfr521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146356.2 | NP_666468.2 | 1 | 518 | ||
GenBank mRNA | BC106825.1 | - | 1 | 516 | ||
BC106826.1 | - | 1 | 518 | 40044016 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146356.2 | NP_666468.2 | 1 | 518 | ||
GenBank mRNA | BC106825.1 | - | 1 | 516 | ||
BC106826.1 | - | 1 | 518 | 40044016 |