siRNA Details
| Gene Symbol: Olfr677 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146358.1 | NP_666470.1 | 1 | 864 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr677| Gene Symbol: Olfr677 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146358.1 | NP_666470.1 | 1 | 864 | ||