siRNA Details
| Gene Symbol: Olfr1312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146362.1 | NP_666474.1 | 1 | 48 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1312| Gene Symbol: Olfr1312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146362.1 | NP_666474.1 | 1 | 48 | ||